Anti-CD32 Mouse Monoclonal Antibody [clone: 7.3]
ANTIA251715-100
New Product
- Antibody type:Primary
- Antigen name:CD32
- Antigen symbol:CD32
- Clonality:Monoclonal
- Clone:7.3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P12318
- Antigen synonyms:Fc fragment of IgG low affinity IIb receptor|CD32C|CD32A|Fc fragment of IgG low affinity IIb receptor for (CD32)|Fc fragment of IgG low affinity IIa receptor for (CD32)|CDw32|Fc fragment of IgG low affinity IIc receptor|CD32B|Fc fragment of IgG low affinity IIa receptor
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:40 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:K562 and FcgRII+L cells
- Tested applications:Functional studies
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [7.3] antibody to CD32 for Functional Studies and IF with samples derived from Human.
- Validated applications: Functional Studies, IF
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: CD32
Clonality: Monoclonal
Clone: 7.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human