Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD34 Rabbit Recombinant Antibody [clone: HPCA1/2598R]
Anti-CD34 Rabbit Recombinant Antibody [clone: HPCA1/2598R]
Catalog # ANTIA250684-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD34 Rabbit Recombinant Antibody [clone: HPCA1/2598R]
Catalog # ANTIA250684-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD34
  • Antigen symbol:
    CD34
  • Clonality:
    Recombinant
  • Clone:
    HPCA1/2598R
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28906
  • Antigen synonyms:
    CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    90 - 110 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human HPCA1 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [HPCA1/2598R] antibody to CD34 for Flow Cytometry and IHC-P with samples derived from Human and Rat.

  • Validated applications: Flow cytometry, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD34
Clonality: Recombinant
Clone: HPCA1/2598R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat