Anti-Calprotectin Mouse Monoclonal Antibody [clone: 47-8D3]
ANTIA253111-100
New Product
- Antibody type:Primary
- Antigen symbol:Calprotectin
- Clonality:Monoclonal
- Clone:47-8D3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat
- Form:Liquid
- Gene ID:UniprotID# P06702
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:14 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human peripheral blood monocyte components
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [47-8D3] antibody to Calprotectin for Flow Cytometry, IF and IHC-P with samples derived from Human and Rat.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Calprotectin
Clonality: Monoclonal
Clone: 47-8D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat