Anti-CD45RB Rabbit Recombinant Antibody [clone: PTPRC/1783R]
Catalog # ANTIA252996-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:B220
- Antigen symbol:CD45RB
- Clonality:Recombinant
- Clone:PTPRC/1783R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P08575
- Antigen synonyms:CD45R|CD45|CD45RB|GP180|Leukocyte common antigen|L-CA|LCA|CD45 antigen|CD 45
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:180 - 220 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CD45RB protein
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [PTPRC/1783R] antibody to CD45RB for Flow Cytometry, IF, WB and IHC-P with samples derived from Human.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD45RB
Clonality: Recombinant
Clone: PTPRC/1783R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human