Anti-p27 KIP 1 Mouse Monoclonal Antibody [clone: DCS-72.F6]
Catalog # ANTIA251280-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AA408329
- Antigen symbol:p27 KIP 1
- Clonality:Monoclonal
- Clone:DCS-72.F6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P46527
- Antigen synonyms:CDKN 1B|CDKN1B|CDKN4|Cyclin dependent kinase inhibitor p27|AI843786|Cyclin Dependent Kinase Inhibitor 1B|CDKN 4|CDN1B_HUMAN|Cdki1b
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:25 - 26 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length mouse p27 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [DCS-72.F6] antibody to p27 KIP 1 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human, Monkey, Mouse and Rat.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 0,25 to 0,5 µg/ml
Type: Primary
Antigen: p27 KIP 1
Clonality: Monoclonal
Clone: DCS-72.F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Mouse, Rat