Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Connexin 32 Rat Monoclonal Antibody [clone: R5.21C]
Catalog # ANTIA250833-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Connexin 32 Rat Monoclonal Antibody [clone: R5.21C]
Catalog # ANTIA250833-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Connexin 32
  • Clonality:
    Monoclonal
  • Clone:
    R5.21C
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Rat,
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28230
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    27 - 32 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Mouse liver DOC-JR-plasma membranes
  • Purification:
    Protein G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rat monoclonal [R5.21C] antibody to Connexin 32 for ELISA with samples derived from Mouse and Rat.

  • Validated applications: ELISA
  • Recommended dilutions: ELISA: 1 to 5 µg/ml

Type: Primary
Antigen: Connexin 32
Clonality: Monoclonal
Clone: R5.21C
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse, Rat