Anti-CD5 Mouse Monoclonal Antibody [clone: B-B8]
Catalog # ANTIA250533-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 5
- Antigen symbol:CD5
- Clonality:Monoclonal
- Clone:B-B8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P06127
- Antigen synonyms:CD5|CD5 molecule|CD5 antigen (p56 62)|CD5 antigen|LEU1|OTTHUMP00000236973|CD5_HUMAN|Lymphocyte antigen T1/Leu-1|LyA
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:67 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:T cells activated with PHA
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [B-B8] antibody to CD5 for Flow Cytometry and IF with samples derived from Human.
- Validated applications: Flow cytometry, IF
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: B-B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human