Anti-alpha 2a Adrenergic Receptor Rabbit Polyclonal Antibody
Catalog # ANTIA11568-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:ADA2A_HUMAN
- Antigen symbol:alpha 2a Adrenergic Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P08913
- Antigen synonyms:Alpha-2 adrenergic receptor subtype C10|ADRAR|Adrenergic alpha 2A receptor|Alpha 2 adrenergic receptor subtype C10|alpha 2a Adrenergic Receptor|Alpha 2AAR subtype C10|ADRA2|ADRA2R|ADRA2A
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
- Molecular weight:49 kDa
- Sequence:RRTRVPPSRRGPDAVAAPPGGTERRPNGLGPERSAGPGGAEAEPLPTQLNGAPGEPAPAGPRDTDALDLEESSSSDHAERPPGPRRPERGPRGKGKARASQVKPGDSLPRRGPGATGIGTPAAGPGEERVGAAKASRWRGR
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 240 - 380 of human ADRA2A (NP_000672.3)
- Tested applications:ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to alpha 2a Adrenergic Receptor for WB and ICC/IF with samples derived from human, mouse and rat.
- Validated applications: WB, ICC/IF
- Recommended dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: alpha 2a Adrenergic Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat