Anti-CLDN16/Claudin-16 Rabbit Polyclonal Antibody
Catalog # ANTIA10818-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:CLDN16/Claudin-16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9Y5I7
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
- Molecular weight:30 kDa
- Sequence:HLSGARAGVCPCCHPDGLLATMRDLLQYIACFFAFFSAGFLIVATWTDCWMVNADDSLEVSTKCRGLWWECVTNAFDGIRTCDEYDSILAEHPLKLVVTRA
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 50 - 150 of human CLDN16 (NP_006571.1)
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to CLDN16 / Claudin-16 for WB with samples derived from human and mouse.
- Validated applications: WB
- Recommended dilutions: WB: 1:100 to 1:500
Type: Primary
Antigen: CLDN16/Claudin-16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse