Anti-CD154 Mouse Monoclonal Antibody [clone: 24-31] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA122116-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 40L
- Antigen symbol:CD154
- Clonality:Monoclonal
- Clone:24-31
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P27548
- Antigen synonyms:CD40 antigen ligand|CD40L|gp39|CD154|CD40-L|CD40LG|CD40L_HUMAN|CD40 ligand, soluble form|CD40 ligand
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human CD154 fusion protein
- Purification:This antibody is conjugated with FITC under optimum conditions The conjugate is purified by size-exclusion chromatography
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [24-31] antibody to CD154 (FITC) for Flow Cytometry with samples derived from human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD154
Clonality: Monoclonal
Clone: 24-31
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Non-Human Primates