Anti-CD64 Mouse Monoclonal Antibody [clone: 10.1] (PE-Cyanine 5)
Catalog # ANTIA122068-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 64
- Antigen symbol:CD64
- Clonality:Monoclonal
- Clone:10.1
- Conjugation:PE (Phycoerythrin)-Cy5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P12314
- Antigen synonyms:Fc fragment of IgG high affinity Ia receptor|Fc fragment of IgG high affinity Ib receptor|CD64c|CD64 antigen|Fc fragment of IgG high affinity Ic receptor|CD64|Fc fragment of IgG, high affinity Ia, receptor (CD64)|CD64A|CD64b
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Rheumatoid synovial fluid cells and fibronectin purified human monocytes
- Purification:This antibody is conjugated with tandem dye PE-Cyanine 5 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [10.1] antibody to CD64 (PE-Cyanine 5) for Flow Cytometry with samples derived from human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: 10.1
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Non-Human Primates