Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CD45 Mouse Monoclonal Antibody [clone: HI30] (PerCP-Cyanine 5.5)
  ANTIA121953-100
New Product
undefined
Anti-CD45 Mouse Monoclonal Antibody [clone: HI30] (PerCP-Cyanine 5.5)
  ANTIA121953-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    B220
  • Antigen symbol:
    CD45
  • Clonality:
    Monoclonal
  • Clone:
    HI30
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5,5®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08575
  • Antigen synonyms:
    CD45R|CD45|GP180|L-CA|Leukocyte Common antigen|LCA|loc|CD45 antigen|CD 45
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Unfortunately, this information is considered to be co mMercially sensitive
  • Purification:
    This antibody is conjugated with tandem dye PerCP-Cyanine 55 under optimum conditions The conjugate is purified by size-exclusion chromatography
  • Pack type:
    Glass bottle
  • Pk:
    100 T

 

 

Mouse monoclonal [HI30] antibody to CD45 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human and Non-Human Primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: HI30
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Non-Human Primates