Anti-CD13 Mouse Monoclonal Antibody [clone: WM15] (PerCP-Cyanine 5.5)
ANTIA121926-100
New Product
- Antibody type:Primary
- Antigen name:Alanyl (membrane) aminopeptidase
- Antigen symbol:CD13
- Clonality:Monoclonal
- Clone:WM15
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5,5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P15144
- Antigen synonyms:ANPEP|AP-N|AP M|AMPN_HUMAN|AP N|Aminopeptidase N|Aminopeptidase M|Alanyl aminopeptidase|AP-M
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human AML cells
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 55 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [WM15] antibody to CD13 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: WM15
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Non-Human Primates