Anti-CD11a Mouse Monoclonal Antibody [clone: MEM-83] (PE (Phycoerythrin))
Catalog # ANTIA242889-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen CD11A
- Antigen symbol:CD11a
- Clonality:Monoclonal
- Clone:MEM-83
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P20701
- Antigen synonyms:alpha polypeptide)|CD 11a|CD11a antigen|Integrin, alpha L (antigen CD11A (p180), lymphocyte function associated antigen 1|Integrin gene promoter|Antigen CD11A (p180), lymphocyte function associated antigen 1, alpha polypeptide|Integrin alpha-L|CD11 antigen-like family member A|Integrin Alpha L
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human peripheral blood lymphocytes
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [MEM-83] antibody to CD11a (PE) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone: MEM-83
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human