Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Ki67 Mouse Monoclonal Antibody [clone: Ki-67] (PE-Cyanine 7)
Anti-Ki67 Mouse Monoclonal Antibody [clone: Ki-67] (PE-Cyanine 7)
Catalog # ANTIA122037-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Ki67 Mouse Monoclonal Antibody [clone: Ki-67] (PE-Cyanine 7)
Catalog # ANTIA122037-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen identified by monoclonal
  • Antigen symbol:
    Ki67
  • Clonality:
    Monoclonal
  • Clone:
    Ki-67
  • Conjugation:
    PE-Cyanine 7
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Bovine
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P46013
  • Antigen synonyms:
    MIB 1|MIB|Antigen KI-67|MKI67|KIA|Marker of proliferation Ki-67|KI67_HUMAN|Antigen KI67|PPP1R105
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Nuclei of the Hodgkin lymphoma cell line L428
  • Purification:
    This antibody is conjugated with tandem dye PE-Cyanine 7 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [Ki-67] antibody to Ki67 (PE-Cyanine 7) for Flow Cytometry with samples derived from human and Bovine.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: Ki67
Clonality: Monoclonal
Clone: Ki-67
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine