Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HLA F Mouse Monoclonal Antibody [clone: 3D11] (PE (Phycoerythrin))
Anti-HLA F Mouse Monoclonal Antibody [clone: 3D11] (PE (Phycoerythrin))
Catalog # ANTIA121831-100
Supplier:  Antibodies.com
New Product
Anti-HLA F Mouse Monoclonal Antibody [clone: 3D11] (PE (Phycoerythrin))
Catalog # ANTIA121831-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CDA12
  • Antigen symbol:
    HLA F
  • Clonality:
    Monoclonal
  • Clone:
    3D11
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P30511
  • Antigen synonyms:
    HLA F antigen|HLA 5.4|HLA class I molecule|HLA-CDA12|HLAF_HUMAN|HLA class I histocompatibility antigen, alpha chain F|HLA-F|HLA F|HLAF
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Concentration:
    0,1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Inclusion body-derived HLA F heavy chain
  • Purification:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [3D11] antibody to HLA F (PE) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1 to 5 µg/ml

Type: Primary
Antigen: HLA F
Clonality: Monoclonal
Clone: 3D11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human