Anti-Prion Protein PrP Mouse Monoclonal Antibody [clone: EM-21]
Catalog # ANTIA121785-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Alternative prion protein
- Antigen symbol:Prion Protein PrP
- Clonality:Monoclonal
- Clone:EM-21
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P04156
- Antigen synonyms:CD230 antigen|ASCR|KURU|AltPrP|p27 30|major prion protein|CD230|GSS|CJD
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human prion protein
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [EM-21] antibody to Prion Protein PrP for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 0,5 µg/ml
Type: Primary
Antigen: Prion Protein PrP
Clonality: Monoclonal
Clone: EM-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human