Anti-GCDFP 15 Rabbit Recombinant Antibody [clone: PIP/9076R]
Catalog # ANTIA316384-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:BRST-2
- Antigen symbol:GCDFP 15
- Clonality:Recombinant
- Clone:PIP/9076R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P12273
- Antigen synonyms:GCDFP-15|Gross cystic disease fluid protein 15|PIP|GPIP4|GCDGP15|GCDFP 15|BRST2|PIP_HUMAN|gp17
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:15 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-146) of human GCDFP-15 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [PIP/9076R] antibody to GCDFP 15 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: GCDFP 15
Clonality: Recombinant
Clone: PIP/9076R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human