Anti-Cytokeratin 17 Rabbit Recombinant Antibody [clone: KRT17/8320R]
Catalog # ANTIA316242-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin 17
- Antigen symbol:Cytokeratin 17
- Clonality:Recombinant
- Clone:KRT17/8320R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q04695
- Antigen synonyms:keratin 17 epitope S2|CK-17|Cytokeratin 17|Keratin|39.1|K1C17_HUMAN|keratin 17 epitope S1|Cytokeratin-17|K17|CK 17
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:46 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment of human KRT17 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [KRT17/8320R] antibody to cytokeratin 17 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: Cytokeratin 17
Clonality: Recombinant
Clone: KRT17/8320R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human