Anti-CD5 Rabbit Recombinant Antibody [clone: C5/8117R]
Catalog # ANTIA316106-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 5
- Antigen symbol:CD5
- Clonality:Recombinant
- Clone:C5/8117R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P06127
- Antigen synonyms:CD5|CD5 molecule|CD5 antigen (p56 62)|CD5 antigen|LEU1|OTTHUMP00000236973|CD5_HUMAN|Lymphocyte antigen T1/Leu-1|LyA
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:67 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant protein corresponding to human extracellular domain of CD5. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [C5/8117R] antibody to CD5 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CD5
Clonality: Recombinant
Clone: C5/8117R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human