Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Catalase Mouse Monoclonal Antibody [clone: HPYL/7227]
Anti-Catalase Mouse Monoclonal Antibody [clone: HPYL/7227]
Catalog # ANTIA315946-100
Supplier:  ANTIBODIES.COM
New Product
Anti-Catalase Mouse Monoclonal Antibody [clone: HPYL/7227]
Catalog # ANTIA315946-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cas1
  • Antigen symbol:
    Catalase
  • Clonality:
    Monoclonal
  • Clone:
    HPYL/7227
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    H. pylori
  • Form:
    Liquid
  • Antigen synonyms:
    Catalase|MGC138422|EC 1.11.1.6|Cs1|CAT|MGC138424|CATA_HUMAN
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    58 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant Helicobacter pylori Catalase protein fragment (around Amino Acids 323-445). The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [HPYL/7227] antibody to Catalase for IHC-P with samples derived from helicobacter pylori.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: Catalase
Clonality: Monoclonal
Clone: HPYL/7227
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Helicobacter pylori