Anti-Keratin 77 Rabbit Recombinant Antibody [clone: KRTL/7784R]
Catalog # ANTIA315212-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Keratin 77
- Clonality:Recombinant
- Clone:KRTL/7784R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q7Z794
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:60 kDa/53 kDa/51 kDa/45 kDa/40 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Type I Cytokeratins residues within aa100-200 of Type I Cytokeratins
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [KRTL/7784R] antibody to Keratin 77 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Keratin 77
Clonality: Recombinant
Clone: KRTL/7784R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human