Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD45 Mouse Monoclonal Antibody [clone: 2B11 + PD7/26]
Anti-CD45 Mouse Monoclonal Antibody [clone: 2B11 + PD7/26]
  ANTIA249809-100
New Product
undefined
Anti-CD45 Mouse Monoclonal Antibody [clone: 2B11 + PD7/26]
  ANTIA249809-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    B220
  • Antigen symbol:
    CD45
  • Clonality:
    Monoclonal
  • Clone:
    2B11 + PD7/26
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    Multiple
  • Reactivity:
    Human,
    Dog
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08575
  • Antigen synonyms:
    CD45R|CD45|GP180|L-CA|Leukocyte Common antigen|LCA|loc|CD45 antigen|CD 45
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    180 - 220 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Clone 2B11: Isolated neoplastic cells from T cell lymphoma. Clone PD7/26: Human peripheral blood lymphocytes maintained in T cell growth factor.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [2B11 + PD7/26] antibody to CD45 for flow cytometry, WB and IHC-P with samples derived from human and canine.

  • Validated applications: Flow cytometry, WB, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 2B11 + PD7/26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human, Dog