Anti-CD45 Mouse Recombinant Antibody [clone: rPTPRC/1461]
Catalog # ANTIA249803-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:B220
- Antigen symbol:CD45
- Clonality:Recombinant
- Clone:rPTPRC/1461
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08575
- Antigen synonyms:CD45R|CD45|GP180|L-CA|Leukocyte Common antigen|LCA|loc|CD45 antigen|CD 45
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:180 - 220 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human PTPRC protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rPTPRC/1461] antibody to CD45 for flow cytometry, IF and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD45
Clonality: Recombinant
Clone: rPTPRC/1461
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human