Anti-Cytokeratin 19 Mouse Recombinant Antibody [clone: rKRT19/799]
ANTIA249207-100
New Product
- Antibody type:Primary
- Antigen name:40 kDa keratin intermediate filament
- Antigen symbol:Cytokeratin 19
- Clonality:Recombinant
- Clone:rKRT19/799
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P08727
- Antigen synonyms:Cytokeratin-19|CK19|CK-19|K1CS|Keratin type I 40kD|Cytokeratin 19|CK 19|Keratin type I 40 kD|K1C19_HUMAN
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:40 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human KRT19 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant mouse monoclonal [rKRT19/799] antibody to Cytokeratin 19 for flow cytometry, IF, WB and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 19
Clonality: Recombinant
Clone: rKRT19/799
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human