Anti-Cytokeratin 17 Mouse Monoclonal Antibody [clone: E3]
ANTIA249189-100
New Product
- Antibody type:Primary
- Antigen name:keratin 17 epitope S2
- Antigen symbol:Cytokeratin 17
- Clonality:Monoclonal
- Clone:E3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Pig,Goat,Human,Bovine,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q04695
- Antigen synonyms:CK-17|Cytokeratin 17|Keratin|39.1|K1C17_HUMAN|keratin 17 epitope S1|Cytokeratin-17|K17|CK 17
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:46 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Cytoskeletal fraction of rat colon epithelium
- Tested applications:IHC
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [E3] antibody to Cytokeratin 17 for flow cytometry, IF, WB and IHC with samples derived from human, rat, bovine, goat and porcine.
- Validated applications: Flow cytometry, IF, WB, IHC
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 17
Clonality: Monoclonal
Clone: E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Rat, Bovine, Goat, Porcine