Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Cytokeratin 15 Rabbit Recombinant Antibody [clone: KRT15/2103R]
Anti-Cytokeratin 15 Rabbit Recombinant Antibody [clone: KRT15/2103R]
Katalogové číslo ANTIA249183-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Cytokeratin 15 Rabbit Recombinant Antibody [clone: KRT15/2103R]
Katalogové číslo ANTIA249183-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytokeratin 15
  • Clonality:
    Recombinant
  • Clone:
    KRT15/2103R
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Bovine,
    Rat
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P19012
  • Antigen synonyms:
    Ka15|CK 15|K1C15_HUMAN|K1CO|AI528832|Cytokeratin15|CK15|CK-15|Cytokeratin-15
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    52 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human KRT15 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [KRT15/2103R] antibody to Cytokeratin 15 for flow cytometry and IHC-P with samples derived from human, rat and bovine.

  • Validated applications: Flow cytometry, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 15
Clonality: Recombinant
Clone: KRT15/2103R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Bovine