Anti-Cytokeratin 6A Mouse Recombinant Antibody [clone: rKRT6A/2100]
ANTIA249131-100
New Product
- Antibody type:Primary
- Antigen name:CK-6A
- Antigen symbol:Cytokeratin 6A
- Clonality:Recombinant
- Clone:rKRT6A/2100
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P02538
- Antigen synonyms:K2C6A_HUMAN|Cytokeratin 6a|Keratin|CK6A|CK6C|Cytokeratin-6A|Cytokeratin-6D|CK-6D|CK6D
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:58 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human Cytokeratin 6A protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant mouse monoclonal [rKRT6A/2100] antibody to Cytokeratin 6A for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 6A
Clonality: Recombinant
Clone: rKRT6A/2100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human