Anti-Galectin 13 Mouse Monoclonal Antibody [clone: PP13/1164]
Katalogové číslo ANTIA248764-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:GAL 13
- Antigen symbol:Galectin 13
- Clonality:Monoclonal
- Clone:PP13/1164
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q9UHV8
- Antigen synonyms:LGALS 13|Galactoside-binding soluble lectin 13|Galectin 13|Gal-13|Galectin-13|Galactoside binding soluble lectin 13|Lectin galactoside binding soluble 13|Galectin13|GAL13
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:16 kDa (monomer)/32 kDa (homodimer)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 23-134, of human Galectin 13 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PP13/1164] antibody to Galectin 13 for ELISA, flow cytometry, IF and IHC-P with samples derived from human.
- Validated applications: ELISA, Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Galectin 13
Clonality: Monoclonal
Clone: PP13/1164
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human