Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD32 Mouse Monoclonal Antibody [clone: 8.7]
Anti-CD32 Mouse Monoclonal Antibody [clone: 8.7]
Catalog # ANTIA248534-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD32 Mouse Monoclonal Antibody [clone: 8.7]
Catalog # ANTIA248534-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    CD32
  • Clonality:
    Monoclonal
  • Clone:
    8.7
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P12318
  • Antigen synonyms:
    Fc fragment of IgG low affinity IIb receptor|CD32C|CD32A|Fc fragment of IgG low affinity IIb receptor for (CD32)|Fc fragment of IgG low affinity IIa receptor for (CD32)|CDw32|Fc fragment of IgG low affinity IIc receptor|CD32B|Fc fragment of IgG low affinity IIa receptor
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    40 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    K562 and FcgRII+L cells
  • Tested applications:
    Functional studies
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [8.7] antibody to CD32 for functional studies and IF with samples derived from human.

  • Validated applications: Functional studies, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: CD32
Clonality: Monoclonal
Clone: 8.7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human