Order Entry
Czech Republic
ContactUsLinkComponent
Anti-EGFR Rabbit Recombinant Antibody [clone: GFR/2968R]
Anti-EGFR Rabbit Recombinant Antibody [clone: GFR/2968R]
Catalog # ANTIA248410-100
Supplier:  Antibodies.com
New Product
undefined
Anti-EGFR Rabbit Recombinant Antibody [clone: GFR/2968R]
Catalog # ANTIA248410-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    9030024J15Rik
  • Antigen symbol:
    EGFR
  • Clonality:
    Recombinant
  • Clone:
    GFR/2968R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P00533
  • Antigen synonyms:
    EGFR_HUMAN|EGF R|EGFR|Avian erythroblastic leukemia viral (verbb) oncogene homolog|Cell growth inhibiting protein 40|AI552599|Epidermal growth factor receptor|Avian erythroblastic leukemia viral (v erb b) oncogene homolog|Cell proliferation inducing protein 61
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    ~170 kDa (wild type)/~145 kDa (vIII variant)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human EGFR protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [GFR/2968R] antibody to EGFR for WB and IHC-P with samples derived from human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: EGFR
Clonality: Recombinant
Clone: GFR/2968R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human