Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Semaphorin 4D Mouse Monoclonal Antibody [clone: SEMA4D/933]
Anti-Semaphorin 4D Mouse Monoclonal Antibody [clone: SEMA4D/933]
Catalog # ANTIA248146-100
Supplier:  Antibodies.com
New Product
Anti-Semaphorin 4D Mouse Monoclonal Antibody [clone: SEMA4D/933]
Catalog # ANTIA248146-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Collapsin4
  • Antigen symbol:
    Semaphorin 4D
  • Clonality:
    Monoclonal
  • Clone:
    SEMA4D/933
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q92854
  • Antigen synonyms:
    CD100 antigen|BB18|Coll 4|BB 18|CD 100|Collapsin 4|COLL4|CD100|A8
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    50 kDa (monomer)
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human SEMA4D protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [SEMA4D/933] antibody to Semaphorin 4D for flow cytometry and IF with samples derived from human and non-human primates.

  • Validated applications: Flow cytometry, IF
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: Semaphorin 4D
Clonality: Monoclonal
Clone: SEMA4D/933
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates