Anti-Semaphorin 4D Mouse Monoclonal Antibody [clone: A8]
Catalog # ANTIA248145-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:A8
- Antigen symbol:Semaphorin 4D
- Clonality:Monoclonal
- Clone:A8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Monkey,Human,Mouse
- Form:Liquid
- Gene ID:UniprotID# Q92854
- Antigen synonyms:CD100 antigen|BB18|Coll 4|BB 18|Collapsin4|CD 100|Collapsin 4|COLL4|CD100
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:50 kDa (monomer)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:PHA stimulated human peripheral blood lymphocytes
- Tested applications:Functional studies
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [A8] antibody to Semaphorin 4D for functional studies, flow cytometry and IF with samples derived from human, monkey and mouse.
- Validated applications: Functional studies, Flow cytometry, IF
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: Semaphorin 4D
Clonality: Monoclonal
Clone: A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Mouse