Order Entry
Czech Republic
ContactUsLinkComponent
Anti-p27 KIP 1 Mouse Monoclonal Antibody [clone: DCS-72.F6]
Anti-p27 KIP 1 Mouse Monoclonal Antibody [clone: DCS-72.F6]
  ANTIA248097-100
New Product
undefined
Anti-p27 KIP 1 Mouse Monoclonal Antibody [clone: DCS-72.F6]
  ANTIA248097-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    AA408329
  • Antigen symbol:
    p27 KIP 1
  • Clonality:
    Monoclonal
  • Clone:
    DCS-72.F6
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Monkey,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P46527
  • Antigen synonyms:
    CDKN 1B|CDKN1B|CDKN4|Cyclin dependent kinase inhibitor p27|AI843786|Cyclin Dependent Kinase Inhibitor 1B|CDKN 4|CDN1B_HUMAN|Cdki1b
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    25 - 26 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length mouse p27 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [DCS-72.F6] antibody to p27 KIP 1 for flow cytometry, IF, WB and IHC-P with samples derived from human, monkey, mouse and rat.

  • Validated applications: Flow cytometry, IF, WB, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 0,25 to 0,5 µg/ml

Type: Primary
Antigen: p27 KIP 1
Clonality: Monoclonal
Clone: DCS-72.F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Mouse, Rat