Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Cadherin 16 Rabbit Recombinant Antibody [clone: CDH16/1532R]
Anti-Cadherin 16 Rabbit Recombinant Antibody [clone: CDH16/1532R]
Catalog # ANTIA248070-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Cadherin 16 Rabbit Recombinant Antibody [clone: CDH16/1532R]
Catalog # ANTIA248070-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CAD16_HUMAN
  • Antigen symbol:
    Cadherin 16
  • Clonality:
    Recombinant
  • Clone:
    CDH16/1532R
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O75309
  • Antigen synonyms:
    CDH16|Cadherin-16|CDH 16|Kidney specific cadherin|Cadherin 16 KSP cadherin|Cadherin 16|Kidney cadherin|Cadherin kidney|Kidney-specific cadherin
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    130 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human CDH16 protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [CDH16/1532R] antibody to Cadherin 16 for WB, flow cytometry and IHC-P with samples derived from human, mouse and rat.

  • Validated applications: WB, Flow cytometry, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, Flow cytometry: 1 to 2 µg/million cells, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cadherin 16
Clonality: Recombinant
Clone: CDH16/1532R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat