Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SHANK2 Mouse Monoclonal Antibody [clone: N23b/49]
Anti-SHANK2 Mouse Monoclonal Antibody [clone: N23b/49]
Catalog # ANTIA304967-100
Supplier:  Antibodies.com
New Product
undefined
Anti-SHANK2 Mouse Monoclonal Antibody [clone: N23b/49]
Catalog # ANTIA304967-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    AUTS17
  • Antigen symbol:
    SHANK2
  • Clonality:
    Monoclonal
  • Clone:
    N23b/49
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9QX74
  • Antigen synonyms:
    KIAA1022|Cortactin SH3 domain-binding protein|CTTNBP1|GKAP/SAPAP-interacting protein|GKAP/SAPAP interacting protein|CortBP1|Proline rich synapse associated protein 1|Cortactin-binding protein 1|Cortactin binding protein 1
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    ~160 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Fusion protein amino acids 84 - 309 of rat Shank2.
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Protein G purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [N23b/49] antibody to SHANK2 for WB, IHC, IP, ICC/IF and antibody microarray with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, IP, ICC/IF, Antibody microarray
  • Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: SHANK2
Clonality: Monoclonal
Clone: N23b/49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat