Anti-SHANK1 Mouse Monoclonal Antibody [clone: N22/21]
ANTIA304815-100
New Product
- Antibody type:Primary
- Antigen name:GKAP/SAPAP interacting protein
- Antigen symbol:SHANK1
- Clonality:Monoclonal
- Clone:N22/21
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9WV48
- Antigen synonyms:SHANK 1|SH3 and multiple ankyrin repeat domains 1|SHANK1|Shank1a|SHAN1_HUMAN|Somatostatin receptor interacting protein|SH3/ankyrin domain gene 1|OTTHUMP00000174437|SH3 and multiple ankyrin repeat domains protein 1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
- Molecular weight:~190 - 220 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Fusion protein amino acids 469 - 691 (SH3/PDZ domains) of rat Shank1.
- Tested applications:IHC
- Purification:Protein G purification
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [N22/21] antibody to SHANK1 for WB, IHC and antibody microarray with samples derived from human, mouse and rat.
- Validated applications: WB, IHC, Antibody microarray
- Recommended dilutions: WB: 1:1000, IHC: 1:1000, ICC/IF: 1:100
Type: Primary
Antigen: SHANK1
Clonality: Monoclonal
Clone: N22/21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat