Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Neuroligin 1 Mouse Monoclonal Antibody [clone: S97A-31]
Anti-Neuroligin 1 Mouse Monoclonal Antibody [clone: S97A-31]
Catalog # ANTIA304803-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Neuroligin 1 Mouse Monoclonal Antibody [clone: S97A-31]
Catalog # ANTIA304803-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    KIAA1070
  • Antigen symbol:
    Neuroligin 1
  • Clonality:
    Monoclonal
  • Clone:
    S97A-31
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q62765
  • Antigen synonyms:
    NLGN1_HUMAN|Nlgn1|NLG 1|NL1|Neuroligin 1|MGC45115|Neuroligin-1|NLG1
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,1% sodium azid
  • Molecular weight:
    ~120 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Fusion protein amino acids 718 - 843 (cytoplasmic C - terminus) of rat Neuroligin - 1. Mouse: 99% identity (125/126 amino acids identical). Human: 99% identity (125/126 amino acids identical) >40% identity with Neuroligin - 2 and - 3..
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Protein G purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [S97A-31] antibody to Neuroligin 1 for WB, IHC and ICC/IF with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: Neuroligin 1
Clonality: Monoclonal
Clone: S97A-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat