Anti-TMEM16A Rabbit Recombinant Antibody [clone: RM440]
Catalog # ANTIA290859-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:ANO 1
- Antigen symbol:TMEM16A
- Clonality:Recombinant
- Clone:RM440
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q5XXA6
- Antigen synonyms:DOG 1|Discovered on gastrointestinal stromal tumors protein 1|Anoctamin1|Anoctamin 1 calcium activated chloride channel|Anoctamin 1|Anoctamin-1|Ca2+ activated Cl- channel|ANO1|ANO1_HUMAN
- Storage buffer:Supplied in phosphate buffered saline with 50% glycerol, 1% BSA, and 0,09% sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human DOG1 protein.
- Tested applications:IHC
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Recombinant rabbit monoclonal [RM440] antibody to TMEM16A for IHC with samples derived from human.
- Validated applications: IHC
- Recommended dilutions: IHC: 1:100 to 1:400
Type: Primary
Antigen: TMEM16A
Clonality: Recombinant
Clone: RM440
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human