Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MMP9 Mouse Monoclonal Antibody [Clone: S51-82]
Anti-MMP9 Mouse Monoclonal Antibody [Clone: S51-82]
Katalogové číslo ANTIA305026-100
Supplier:  Antibodies.com
New Product
undefined
Anti-MMP9 Mouse Monoclonal Antibody [Clone: S51-82]
Katalogové číslo ANTIA305026-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    82 kDa matrix metalloproteinase-9
  • Antigen symbol:
    MMP9
  • Clonality:
    Monoclonal
  • Clone:
    S51-82
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P50282
  • Antigen synonyms:
    92 kDa gelatinase|92 kDa type IV collagenase|Collagenase Type 4 beta|EC 3.4.24.35|Gelatinase 92 KD|CLG 4B|Gelatinase B|Collagenase type IV 92 KD|CLG4B
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 50% glycerol and 0,09% sodium azide
  • Molecular weight:
    ~82/~92 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Fusion protein amino acids 1 - 708 (full length) of rat MMP9.
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Protein G purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [S51-82] antibody to MMP9 for WB, IHC, ICC/IF, IP and Flow cytometry with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC, ICC/IF, IP, Flow cytometry
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:100

Type: Primary
Antigen: MMP9
Clonality: Monoclonal
Clone: S51-82
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat