Order Entry
Czech Republic
ContactUsLinkComponent
Bovine Thyroxine/T4 ELISA Kit
Bovine Thyroxine/T4 ELISA Kit
Katalogové číslo ANTIA87529-96
Supplier:  Antibodies.com
New Product
CAS Number:  
undefined
Bovine Thyroxine/T4 ELISA Kit
Katalogové číslo ANTIA87529-96
Supplier:  Antibodies.com
New Product
CAS Number:  

Specifications

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Bovine
  • Target protein:
    Thyroxine/T4
  • Description:
    Bovine thyroxine/T4 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates and other biological fluids
  • Cross reactivity:
    Bovine Thyroxine/T4 ELISA Kit exhibits high specificity and excellent specificity for the detection of bovine Thyroxine/T4. No significant cross-reactivity or interference between Thyroxine/T4 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Assay Principle:
    Quantitative
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,703 - 45 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    0,422 ng/ml
  • Regulatory status:
    RUO

Specifications

About this item

Bovine Thyroxine/T4 ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of bovine Thyroxine/T4 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 0,703 to 45 ng/ml
  • Sensitivity: 0,422 ng/ml
  • Sample type: Serum, plasma, tissue homogenates and other biological fluids
  • Assay precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Bovine Thyroxine/T4 ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of bovine Thyroxine/T4 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Thyroxine/T4 antigen. During the incubation, Thyroxine/T4 present in the samples or standards competes with the fixed amount of immobilised Thyroxine/T4 for binding sites on the Biotinylated Anti-Thyroxine/T4 Antibody. The more Thyroxine/T4 present in a sample or standard, the less Biotinylated Anti-Thyroxine/T4 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Thyroxine/T4 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Thyroxine/T4 present in each sample or standard. The concentration of Thyroxine/T4 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.