Order Entry
Czech Republic
ContactUsLinkComponent
 
Human SIRP alpha ELISA Kit
  ANTIA312560-96
New Product
 :  
undefined
Human SIRP alpha ELISA Kit
  ANTIA312560-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    SIRP alpha
  • Description:
    Human SIRP alpha ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,75 - 12 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    0,031 ng/ml
  • Regulatory status:
    RUO

 

 

Human SIRP alpha ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human SIRP alpha in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 0,75 to 12 ng/ml
  • Sensitivity: 0,031 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human SIRP alpha ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human SIRP alpha in serum, plasma or other biological fluids. An antibody specific for SIRP alpha has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SIRP alpha present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-SIRP alpha antibody, which also binds the SIRP alpha present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SIRP alpha antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SIRP alpha antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of SIRP alpha captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of SIRP alpha in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.