Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Human Integrin Beta 7 ELISA Kit

Human Integrin Beta 7 ELISA Kit

Human Integrin beta 7 ELISA kit, 96 Tests
Katalogové číslo ANTIA312352-96
Dodavatel:   Antibodies.com
Human Integrin Beta 7 ELISA Kit
Katalogové číslo ANTIA312352-96
Dodavatel:  Antibodies.com
Číslo CAS:  
Human Integrin Beta 7 ELISA Kit
Human Integrin Beta 7 ELISA Kit

Human Integrin beta 7 ELISA kit, 96 Tests

Přejít na:
Číslo CAS:  
Price
14 130,00 Kč
Přihlásit se pro dostupnost
1 * 96 Tests
Product Details & Documents

Human Integrin beta 7 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Integrin beta 7 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 1,5 to 24 ng/ml
  • Sensitivity: 0,051 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human Integrin beta 7 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Integrin beta 7 in serum, plasma or other biological fluids. An antibody specific for Integrin beta 7 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Integrin beta 7 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Integrin beta 7 antibody, which also binds the Integrin beta 7 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Integrin beta 7 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Integrin beta 7 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Integrin beta 7 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Integrin beta 7 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifikace

Specifikace

  • Pk:
    96 Tests
  • Description:
    Human Integrin beta 7 ELISA kit
  • Primary antibody reactivity:
    Human
  • Sample type:
    Serum, plasma or other biological fluids
  • Sample volume:
    40 μl
  • Detection range:
    1,5 - 24 ng/ml
  • Storage temperature:
    4 °C
  • Shelf life:
    Store for 6 months at 4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ANTIA312352-96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,051 ng/ml
  • Format:
    Pre-coated
  • Target protein:
    Integrin beta 7
  • Regulatory status:
    RUO

Specifikace