Order Entry
Czech Republic
ContactUsLinkComponent
 
Mouse SMPDL3A ELISA Kit
  ANTIA310516-96
New Product
 :  
undefined
Mouse SMPDL3A ELISA Kit
  ANTIA310516-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    SMPDL3A
  • Description:
    Mouse SMPDL3A ELISA kit
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0,75 - 12 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    0,024 ng/ml
  • Regulatory status:
    RUO

 

 

Mouse SMPDL3A ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse SMPDL3A in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 0,75 to 12 ng/ml
  • Sensitivity: 0,024 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse SMPDL3A ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse SMPDL3A in serum, plasma or other biological fluids. An antibody specific for SMPDL3A has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the SMPDL3A present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-SMPDL3A antibody, which also binds the SMPDL3A present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-SMPDL3A antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-SMPDL3A antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of SMPDL3A captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of SMPDL3A in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.