Specifikace
- Pk:96 Tests
- Assay duration:One step
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:ROMO1
- Description:Mouse ROMO1 ELISA kit
- Sample type:Serum, plasma or other biological fluids
- Detection method:Colorimetric
- Time to Results:1 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:75 - 1200 ng/L
- Storage temperature:4 °C
- Sample volume:40 μl
- Sensitivity:5,97 ng/L
- Regulatory status:RUO
Specifikace
O tomto produktu
Mouse ROMO1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse ROMO1 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutes, with a single wash step
- Detection range: 75 to 1200 ng/L
- Sensitivity: 5,97 ng/L
- Sample type: Serum, plasma or other biological fluids
- Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse ROMO1 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse ROMO1 in serum, plasma or other biological fluids. An antibody specific for ROMO1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ROMO1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-ROMO1 antibody, which also binds the ROMO1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-ROMO1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-ROMO1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of ROMO1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of ROMO1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.