- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:C1QB
- Description:Rat C1QB ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Rat C1QB ELISA Kit exhibits high specificity and excellent specificity for the detection of rat C1QB. No significant cross-reactivity or interference between C1QB and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:15,625 - 1000 pg/ml
- Storage temperature:4 °C
- Sensitivity:9,375 pg/ml
- Regulatory status:RUO
Rat C1QB ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat C1QB in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Rat C1QB ELISA kit (A303806) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat C1QB in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for C1QB has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the C1QB present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-C1QB antibody, which binds the captured C1QB present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of C1QB captured in each well. The concentration of C1QB can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.