Rat Ovalbumin Specific IgE ELISA Kit
Catalog # ANTIA303777-96
Supplier: ANTIBODIES.COM
New Product
CAS Number:
Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Indirect
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:Ovalbumin Specific IgE
- Description:Rat ovalbumin specific IgE ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,781 - 50 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,469 ng/ml
- Regulatory status:RUO
Specifications
About this item
Rat Ovalbumin Specific IgE ELISA kit is a capture Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of rat Ovalbumin Specific IgE in serum, plasma, tissue homogenates, and other biological fluids.