Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Rat
- Target protein:EAAT2
- Description:Rat EAAT2 ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Cross reactivity:Rat EAAT2 ELISA Kit exhibits high specificity and excellent specificity for the detection of rat EAAT2. No significant cross-reactivity or interference between EAAT2 and analogues was observed.
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:31,25 - 2000 pg/ml
- Storage temperature:4 °C
- Sensitivity:18,75 pg/ml
- Regulatory status:RUO
Specifications
About this item
Rat EAAT2 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat EAAT2 in serum, plasma, tissue homogenates, and other biological fluids.
- Ready to use ELISA kit
- Detection Range: 31,25 to 2000 pg/ml
- Sensitivity: 18,75 pg/ml
- Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
- Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%
Rat EAAT2 ELISA kit (A303742) employs the sandwich enzyme immunoassay technique for the quantitative measurement of rat EAAT2 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for EAAT2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the EAAT2 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated anti-EAAT2 antibody, which binds the captured EAAT2 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of EAAT2 captured in each well. The concentration of EAAT2 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.