Order Entry
Czech Republic
ContactUsLinkComponent
 
Rat Rheumatoid Factor IgG ELISA Kit
  ANTIA303718-96
New Product
 :  
undefined
Rat Rheumatoid Factor IgG ELISA Kit
  ANTIA303718-96
New Product
 :  

 

  • Pk:
    96 Tests
  • Assay duration:
    Multiple steps
  • Assay Type:
    Indirect
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Rheumatoid Factor IgG
  • Description:
    Rat rheumatoid factor IgG ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    3,125 - 200 IU/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    1,875 IU/ml
  • Regulatory status:
    RUO

 

 

Rat Rheumatoid Factor IgG ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of rat Rheumatoid Factor IgG in serum, plasma, tissue homogenates, and other biological fluids.