Mouse Anti-Beta Amyloid 42 Antibody ELISA Kit
Catalog # ANTIA303492-96
Supplier: Antibodies.com
New Product
CAS Number:
Specifications
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Indirect
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:Anti-beta Amyloid 42 Antibody
- Description:Mouse anti-beta amyloid 42 antibody ELISA kit
- Sample type:Serum, plasma, tissue homogenates, and other biological fluids
- Detection method:Colorimetric
- Time to Results:4 h 30 min
- Assay Principle:Quantitative
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,313 - 20 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,188 ng/ml
- Regulatory status:RUO
Specifications
About this item
Mouse Anti-beta Amyloid 42 Antibody ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Anti-beta Amyloid 42 Antibody in serum, plasma, tissue homogenates, and other biological fluids.